missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 4-vinylbenzoate, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.
Notes
Incompatible with oxidizing agents.
Methyl 4-vinylbenzoate is used as a fluid component for enhancing ink stability in printing and also in preparing vinyl substituted beta-diketones. Further, it is used as an organic intermediate for pharmaceuticals.
Notes
Incompatible with oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 32°C to 37°C |
Quantity | 5 g |
Formula Weight | 162.19 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 4-vinylbenzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'