Learn More
Methyl 4H-furo[3,2-b]pyrrole-5-carboxylate, 97%, Thermo Scientific™
£47.20 - £273.26
Chemical Identifiers
CAS | 77484-99-2 |
---|---|
Molecular Formula | C8H7NO3 |
Molecular Weight (g/mol) | 165.15 |
MDL Number | MFCD06797487 |
InChI Key | LIIJOCBTGRORSY-UHFFFAOYSA-N |
Synonym | methyl 4h-furo 3,2-b pyrrole-5-carboxylate, 4h-furo 3,2-b pyrrole-5-carboxylic acid methyl ester, methyl furano 2,3-d pyrrole-5-carboxylate, 5-methoxycarbonyl-4h-furo 3,2-b pyrrole, 4h-furo 3,2-b pyrrole-5-carboxylicacid, methyl ester, 4h-furo 3,2-b pyrrole-5-carboxylic acid, methyl ester |
PubChem CID | 5082263 |
IUPAC Name | methyl 4H-furo[3,2-b]pyrrole-5-carboxylate |
SMILES | COC(=O)C1=CC2=C(N1)C=CO2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10443413
|
Thermo Scientific Maybridge
CC39222DA |
1 g | Amber Glass Bottle |
£47.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10769573
|
Thermo Scientific Maybridge
CC39222EA |
10 g | Amber Glass Bottle | N/A | |||||
Description
Chemical Identifiers
77484-99-2 | |
165.15 | |
LIIJOCBTGRORSY-UHFFFAOYSA-N | |
5082263 | |
COC(=O)C1=CC2=C(N1)C=CO2 |
C8H7NO3 | |
MFCD06797487 | |
methyl 4h-furo 3,2-b pyrrole-5-carboxylate, 4h-furo 3,2-b pyrrole-5-carboxylic acid methyl ester, methyl furano 2,3-d pyrrole-5-carboxylate, 5-methoxycarbonyl-4h-furo 3,2-b pyrrole, 4h-furo 3,2-b pyrrole-5-carboxylicacid, methyl ester, 4h-furo 3,2-b pyrrole-5-carboxylic acid, methyl ester | |
methyl 4H-furo[3,2-b]pyrrole-5-carboxylate |
Specifications
77484-99-2 | |
100.0 | |
C8H7NO3 | |
methyl 4h-furo 3,2-b pyrrole-5-carboxylate, 4h-furo 3,2-b pyrrole-5-carboxylic acid methyl ester, methyl furano 2,3-d pyrrole-5-carboxylate, 5-methoxycarbonyl-4h-furo 3,2-b pyrrole, 4h-furo 3,2-b pyrrole-5-carboxylicacid, methyl ester, 4h-furo 3,2-b pyrrole-5-carboxylic acid, methyl ester | |
COC(=O)C1=CC2=C(N1)C=CO2 | |
165.15 | |
165.15 | |
Methyl 4H-furo[3, 2-b]pyrrole-5-carboxylate |
96.5 | |
97% | |
MFCD06797487 | |
LIIJOCBTGRORSY-UHFFFAOYSA-N | |
methyl 4H-furo[3,2-b]pyrrole-5-carboxylate | |
5082263 | |
97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
Your input is important to us. Please complete this form to provide feedback related to the content on this product.