missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-chloro-2-methoxybenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11469702
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33924-48-0
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.618
MDL Number MFCD00017511
InChI Key HPTHYBXMNNGQEF-UHFFFAOYSA-N
Synonym methyl 5-chloro-o-anisate, methyl-5-chloro-2-methoxybenzoate, 5-chloro-2-methoxybenzoic acid methyl ester, benzoic acid, 5-chloro-2-methoxy-, methyl ester, 5-chloro-o-anisic acid methyl ester, 5-chloro-2-methoxy-benzoic acid methyl ester, pubchem3709, methyl5-chloro-2-methoxybenzoate, acmc-1agpb, dsstox_cid_31509
PubChem CID 118574
IUPAC Name methyl 5-chloro-2-methoxybenzoate
SMILES COC1=C(C=C(C=C1)Cl)C(=O)OC

Specifications

Density 1.259
Boiling Point 235°C to 240°C
Flash Point >110°C (230°F)
Refractive Index 1.546
Quantity 100 g
Beilstein 2719088
Formula Weight 200.62
Percent Purity 98%
Chemical Name or Material Methyl 5-chloro-2-methoxybenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'