missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-fluoro-2-nitrobenzoate, 98%, Thermo Scientific Chemicals

Product Code. 15457828
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl 5-fluoro-2-nitrobenzoate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 5-fluoro-2-nitrobenzoate is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water. Soluble in dimethyl sulfoxide and acetone.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 393-85-1
Molecular Formula C8H6FNO4
Molecular Weight (g/mol) 199.14
MDL Number MFCD03425619
InChI Key LCAUVFVOLNRVKG-UHFFFAOYSA-N
Synonym 2-nitro-5-fluorobenzoic acid methyl ester, 5-fluoro-2-nitrobenzoic acid methyl ester, 2-nitro-5-fluorobenzoic acid, methyl ester, benzoic acid, 5-fluoro-2-nitro-, methyl ester, methyl 5fluoro-2-nitrobenzoate, methyl5-fluoro-2-nitrobenzoate, 2-nitro-5-fluorobenzoic acid,methyl ester, 3-fluoro-6-nitrobenzoic acid methyl ester, 5-fluoro-2-nitro-benzoic acid methyl ester, benzoic acid,5-fluoro-2-nitro-, methyl ester
PubChem CID 5152049
IUPAC Name methyl 5-fluoro-2-nitrobenzoate
SMILES COC(=O)C1=C(C=CC(F)=C1)[N+]([O-])=O

Specifications

Melting Point 35°C to 37°C
Color White
Quantity 5 g
Solubility Information Slightly soluble in water. Soluble in dimethyl sulfoxide and acetone.
Formula Weight 199.14
Percent Purity 98%
Physical Form Low Melting Solid
Chemical Name or Material Methyl 5-fluoro-2-nitrobenzoate
compliance-icons
Product Identifier
  • Methyl 5-fluoro-2-nitrobenzoate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P363-Wash contaminated clothing before reuse.
  • P403-Store in a well-ventilated place.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Methyl 5-fluoro-2-nitrobenzoate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.