missing translation for 'onlineSavingsMsg'
Learn More

Methyl 5-iodosalicylate, 99%, Thermo Scientific Chemicals

Product Code. 11419547
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4068-75-1
Molecular Formula C8H7IO3
Molecular Weight (g/mol) 278.045
MDL Number MFCD00016462
InChI Key NRSWJTRJHPRZMH-UHFFFAOYSA-N
PubChem CID 248910
IUPAC Name methyl 2-hydroxy-5-iodobenzoate
SMILES COC(=O)C1=C(C=CC(=C1)I)O

Specifications

Melting Point 75°C to 78°C
Quantity 250 g
Sensitivity Light sensitive
Synonym methyl 5-iodosalicylate, 5-iodosalicylic acid methyl ester, 2-hydroxy-5-iodobenzoic acid methyl ester, benzoic acid, 2-hydroxy-5-iodo-, methyl ester, methyl 2-hydroxy-5-iodobenzenecarboxylate, salicylic acid, 5-iodo-, methyl ester, methyl5-iodosalicylate, methyl-5-iodosalicylate, acmc-209jg0, methyl2-hydroxy-5-iodobenzoate
Formula Weight 278.05
Percent Purity 99%
Chemical Name or Material Methyl 5-iodosalicylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'