missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 6-bromoindole-2-carboxylate, 97%, Thermo Scientific Chemicals
POA - £272.00
Chemical Identifiers
CAS | 372089-59-3 |
---|---|
MDL Number | MFCD03094942 |
Synonym | ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester |
PubChem CID | 7009496 |
IUPAC Name | ethyl 6-bromo-1H-indole-2-carboxylate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15419748
|
Thermo Scientific Alfa Aesar
H63187.MD |
250 mg | |||||||
15499738
|
Thermo Scientific Alfa Aesar
H63187.03 |
1 g |
£58.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15409748
|
Thermo Scientific Alfa Aesar
H63187.06 |
5 g |
£272.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl 6-bromoindole-2-carboxylate, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in acetone.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
372089-59-3 | |
ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester | |
ethyl 6-bromo-1H-indole-2-carboxylate |
MFCD03094942 | |
7009496 |
Specifications
372089-59-3 | |
C11H10BrNO2 | |
ethyl 6-bromoindole-2-carboxylate, 6-bromoindole-2-carboxylic acid ethyl ester, 6-bromo-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromo-2-indolecarboxylic ethyl ester, 6-bromo-2-ethoxylcarbonyl-1h-indole, 6-bromo-2-carbethoxyindole, 6-bromo-2-carboethoxyindole, 1h-indole-2-carboxylicacid, 6-bromo-, ethyl ester | |
FVMZWWFKRMBNSZ-UHFFFAOYSA-N | |
ethyl 6-bromo-1H-indole-2-carboxylate | |
7009496 | |
97% | |
Methyl 6-bromoindole-2-carboxylate |
White | |
MFCD03094942 | |
Soluble in acetone. | |
CCOC(=O)C1=CC2=C(N1)C=C(C=C2)Br | |
268.11 | |
254.08 | |
Crystalline Powder |
Safety and Handling
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only