missing translation for 'onlineSavingsMsg'
Learn More

Methyl 7-chloroheptanoate, 98%, Thermo Scientific™

Product Code. 11406180
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26040-62-0
Molecular Formula C8H15ClO2
Molecular Weight (g/mol) 178.66
MDL Number MFCD09751145
InChI Key YKSFGXAJDCLJKE-UHFFFAOYSA-N
Synonym 7-chloroheptanoic acid, methyl ester, heptanoic acid, 7-chloro-, methyl ester, acmc-20akfu, methyl 7-chloroheptanoate, 7-chloroheptanoic acid methyl ester, heptanoic acid, 7-chloro, methyl ester
PubChem CID 529989
IUPAC Name methyl 7-chloroheptanoate
SMILES COC(=O)CCCCCCCl

Specifications

Linear Formula Cl(CH2)6CO2CH3
Refractive Index 1.4425
Quantity 5 g
Formula Weight 178.66
Percent Purity 98%
Chemical Name or Material Methyl 7-chloroheptanoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'