missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Acetate, ReagentPlus™, 99%, Honeywell™
Click to view available options
Quantity:
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -98°C |
Color | Colorless |
Density | 0.934g/mL at 25°C |
Boiling Point | 58°C |
Flash Point | −13°C (8.6°F) |
Packaging | Glass Bottle |
Linear Formula | CH3COOCH3 |
Refractive Index | n20/D 1.361(lit.) |
Quantity | 2.5 L |
UN Number | UN1231 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'