missing translation for 'onlineSavingsMsg'
Learn More

Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10133560
Click to view available options
Quantity:
10 mL
500 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-33-3, 150-76-5
Molecular Formula C4H6O2
Molecular Weight (g/mol) 86.09
MDL Number MFCD00008627
InChI Key BAPJBEWLBFYGME-UHFFFAOYSA-N
Synonym methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene
PubChem CID 7294
ChEBI CHEBI:82482
IUPAC Name methyl prop-2-enoate
SMILES COC(=O)C=C

Specifications

Melting Point -75°C
Density 0.9560g/mL
Boiling Point 80°C
Flash Point -3°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCO2CH3
Refractive Index 1.4010 to 1.404
Quantity 500 mL
Beilstein 02, 399
Fieser 05,439; 13,183; 17,183
Merck Index 15, 6085
Specific Gravity 0.956
Solubility Information Solubility in water: 60g/L (20°C). Other solubilities: soluble in alcohol and ether
Stabilizer 10 to 20ppm MEHQ
Viscosity 0.50 mPa.s (20°C)
Formula Weight 86.09
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl acrylate, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl acrylate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'