Learn More
Methyl acrylate, 99%, stabilized, Thermo Scientific Chemicals
£26.80 - £158.00
Chemical Identifiers
CAS | 96-33-3, 150-76-5 |
---|---|
Molecular Formula | C4H6O2 |
Molecular Weight (g/mol) | 86.09 |
MDL Number | MFCD00008627 |
InChI Key | BAPJBEWLBFYGME-UHFFFAOYSA-N |
Synonym | methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene |
PubChem CID | 7294 |
ChEBI | CHEBI:82482 |
IUPAC Name | methyl prop-2-enoate |
SMILES | COC(=O)C=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10478280
|
Thermo Scientific Acros
126190100 |
10 mL | Glass bottle |
£26.80
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10133560
|
Thermo Scientific Acros
126195000 |
500 mL | Glass bottle |
£26.90
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10713701
|
Thermo Scientific Acros
126190010 |
1 L | Glass bottle |
£31.60
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10091140
|
Thermo Scientific Acros
126190025 |
2.5 L | Glass bottle |
£58.80
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10465555
|
Thermo Scientific Acros
126190101 |
10 L | Metal Drum |
£158.00
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
96-33-3, 150-76-5 | |
86.09 | |
BAPJBEWLBFYGME-UHFFFAOYSA-N | |
7294 | |
methyl prop-2-enoate |
C4H6O2 | |
MFCD00008627 | |
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene | |
CHEBI:82482 | |
COC(=O)C=C |
Specifications
96-33-3, 150-76-5 | |
0.9560g/mL | |
-3°C | |
98.5% min. (GC) | |
H2C=CHCO2CH3 | |
MFCD00008627 | |
05,439; 13,183; 17,183 | |
0.956 | |
Solubility in water: 60g/L (20°C). Other solubilities: soluble in alcohol and ether | |
COC(=O)C=C | |
10 to 20ppm MEHQ | |
7294 | |
0.50 mPa.s (20°C) | |
99% | |
Methyl acrylate, Stabilized |
-75°C | |
80°C | |
Authentic | |
C4H6O2 | |
1.4010 to 1.404 | |
02, 399 | |
15, 6085 | |
methyl acrylate, methylacrylate, acrylic acid methyl ester, 2-propenoic acid, methyl ester, methyl 2-propenoate, methyl propenoate, methylacrylaat, metilacrilato, methyl-acrylat, methoxycarbonylethylene | |
BAPJBEWLBFYGME-UHFFFAOYSA-N | |
methyl prop-2-enoate | |
86.09 | |
CHEBI:82482 | |
86.09 | |
Liquid |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic if inhaled
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Ground/Bond container and receiving equipment.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a positi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-500-6
- Methyl acrylate, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only