missing translation for 'onlineSavingsMsg'
Learn More

Methyl aminomethanimidothioate hydroiodide, 95%, Thermo Scientific™

Product Code. 10095472
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4338-95-8
Molecular Formula C2H7IN2S
Molecular Weight (g/mol) 218.06
MDL Number MFCD00035598
InChI Key LFXAECSQJSRSTP-UHFFFAOYSA-N
Synonym methyl aminomethanimidothioate hydroiodide, sulfiode, methylthiouronium iodide, methyl carbamimidothioate hydroiodide, s-methylthiuronium iodide, s-methylthiouronium iodide, s-methylisothiouronium iodide, 2-methyl-2-thiopseudourea hydroiodide, 2-methylisothiouronium iodide, s-methylisothiourea hydriodide
PubChem CID 197812
SMILES [H+].[I-].CSC(N)=N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl carbamimidothioate;hydroiodide
Formula Weight 218.06
Percent Purity 95%
Chemical Name or Material Methyl aminomethanimidothioate hydroiodide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'