missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl anthranilate, 99%, Thermo Scientific Chemicals
£22.50 - £48.30
Chemical Identifiers
CAS | 134-20-3 |
---|---|
Molecular Formula | C8H9NO2 |
Molecular Weight (g/mol) | 151.17 |
InChI Key | VAMXMNNIEUEQDV-UHFFFAOYSA-N |
PubChem CID | 8635 |
IUPAC Name | methyl 2-aminobenzoate |
SMILES | COC(=O)C1=CC=CC=C1N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11467627
|
Thermo Scientific Alfa Aesar
A15681.30 |
250 g |
£22.50
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11438810
|
Thermo Scientific Alfa Aesar
A15681.36 |
500 g |
£34.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11457627
|
Thermo Scientific Alfa Aesar
A15681.0B |
1000 g |
£48.30
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is applied as a flavoring agent. Food companies use artificial MA in their candy and chewing gum to give them their grape flavor. Beverage companies have been adding it to the carbonated water and sugar in their soft drinks for the same reason.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
134-20-3 | |
151.17 | |
8635 | |
COC(=O)C1=CC=CC=C1N |
C8H9NO2 | |
VAMXMNNIEUEQDV-UHFFFAOYSA-N | |
methyl 2-aminobenzoate |
Specifications
134-20-3 | |
260°C | |
14,6020 | |
VAMXMNNIEUEQDV-UHFFFAOYSA-N | |
methyl 2-aminobenzoate | |
8635 | |
Liquid |
24°C | |
C8H9NO2 | |
1S/C8H9NO2/c1-11-8(10)6-4-2-3-5-7(6)9/h2-5H,9H2,1H3 | |
COC(=O)C1=CC=CC=C1N | |
151.17 | |
99% | |
Methyl anthranilate |