missing translation for 'onlineSavingsMsg'
Learn More

Methyl anthranilate, 99%

Product Code. p-160001 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
2.5kg
250g
500g
50g
This item is not returnable. View return policy

Product Code. 10784071

Brand: Thermo Scientific Acros 126320500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 134-20-3 | C8H9NO2 | 151.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134-20-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007710
InChI Key VAMXMNNIEUEQDV-UHFFFAOYSA-N
Synonym methyl anthranilate, 2-aminobenzoic acid methyl ester, anthranilic acid methyl ester, methyl o-aminobenzoate, nevoli oil, methylanthranilate, o-carbomethoxyaniline, 2-carbomethoxyaniline, 2-methoxycarbonyl aniline, benzoic acid, 2-amino-, methyl ester
PubChem CID 8635
ChEBI CHEBI:73244
IUPAC Name methyl 2-aminobenzoate
SMILES COC(=O)C1=CC=CC=C1N

Specifications

Color Brown-Yellow
Density 1.1680g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula 2-(H2N)C6H4CO2CH3
Refractive Index 1.5810 to 1.5840 (20°C, 589nm)
Quantity 50 g
Beilstein 14, 317
Merck Index 15, 6094
Formula Weight 151.16
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Methyl anthranilate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Methyl anthranilate
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.