missing translation for 'onlineSavingsMsg'
Learn More

Methyl behenate, 99%, analytical standard for GC, Thermo Scientific Chemicals

Product Code. 15590163
Click to view available options
Quantity:
1 g
5 g

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-77-1
Molecular Formula C23H46O2
Molecular Weight (g/mol) 354.62
MDL Number MFCD00009347
InChI Key QSQLTHHMFHEFIY-UHFFFAOYSA-N
Synonym methyl behenate, docosanoic acid, methyl ester, behenic acid methyl ester, docosanoic acid methyl ester, behenic acid, methyl ester, unii-04kbo9r771, behenic methyl ester, kemester 9022, docosanoic acid methyl, acmc-209rjf
PubChem CID 13584
IUPAC Name methyl docosanoate
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)OC

Specifications

Melting Point 54.0°C to 56.0°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)20CO2CH3
Quantity 5 g
Merck Index 15,1022
Solubility Information (5% in Hexane) Clear colorless solution
Formula Weight 354.61
Percent Purity 99%
Chemical Name or Material Methyl behenate
Product Identifier
  • Methyl behenate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'