missing translation for 'onlineSavingsMsg'
Learn More

Methyl biphenyl-4-carboxylate, 98+%, Thermo Scientific Chemicals

Product Code. 11458810
Click to view available options
Quantity:
5 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 720-75-2
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00017200
InChI Key GATUGNVDXMYTJX-UHFFFAOYSA-N
Synonym methyl 4-biphenylcarboxylate, methyl biphenyl-4-carboxylate, methyl 1,1'-biphenyl-4-carboxylate, methyl p-phenylbenzoate, 4-biphenylcarboxylic acid, methyl ester, 1,1'-biphenyl-4-carboxylic acid, methyl ester, p-phenylbenzoic acid methyl ester, methyl-4-phenylbenzoate, 4-phenylbenzoic acid methyl ester
PubChem CID 69757
IUPAC Name methyl 4-phenylbenzoate
SMILES COC(=O)C1=CC=C(C=C1)C1=CC=CC=C1

Specifications

Melting Point 116°C to 119°C
Odor Odorless
Quantity 5 g
Beilstein 1954046
Formula Weight 212.25
Percent Purity ≥98%
Chemical Name or Material Methyl biphenyl-4-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'