missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl cyanoacetate, 97%, Thermo Scientific Chemicals
£27.40 - £69.40
Chemical Identifiers
CAS | 105-34-0 |
---|---|
Molecular Formula | C4H5NO2 |
Molecular Weight (g/mol) | 99.09 |
MDL Number | MFCD00001939 |
InChI Key | ANGDWNBGPBMQHW-UHFFFAOYSA-N |
Synonym | methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate |
PubChem CID | 7747 |
ChEBI | CHEBI:51928 |
IUPAC Name | methyl 2-cyanoacetate |
SMILES | COC(=O)CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10617502
|
Thermo Scientific Acros
148642500 |
250 mL | Glass bottle |
£27.40
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10123150
|
Thermo Scientific Acros
148640010 |
1 L | Glass bottle |
£69.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105-34-0 | |
99.09 | |
ANGDWNBGPBMQHW-UHFFFAOYSA-N | |
7747 | |
methyl 2-cyanoacetate |
C4H5NO2 | |
MFCD00001939 | |
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate | |
CHEBI:51928 | |
COC(=O)CC#N |
Specifications
105-34-0 | |
Colorless to Yellow | |
204°C to 207°C | |
Authentic | |
C4H5NO2 | |
1.4170 to 1.419 | |
02, 584 | |
methyl cyanoacetate, cyanoacetic acid methyl ester, methyl cyanoethanoate, acetic acid, cyano-, methyl ester, usaf kf-22, methylester kyseliny kyanoctove, methylcyanoacetate, malonic methyl ester nitrile, acetic acid, 2-cyano-, methyl ester, methyl cyano acetate | |
ANGDWNBGPBMQHW-UHFFFAOYSA-N | |
methyl 2-cyanoacetate | |
7747 | |
99.09 | |
Liquid |
-13°C | |
1.1200g/mL | |
>110°C | |
96% min. (GC) | |
NCCH2CO2CH3 | |
MFCD00001939 | |
1.12 | |
Solubility in water: 54g/L in water (20°C). Other solubilities: soluble in alcohol and ether | |
COC(=O)CC#N | |
99.09 | |
CHEBI:51928 | |
97% | |
Methyl cyanoacetate, 97% |
Safety and Handling
missing translation for 'einecsNumber' : 203-288-8
missing translation for 'rtecsNumber' : AG4375000
missing translation for 'tsca' : TSCA
RUO – Research Use Only