missing translation for 'onlineSavingsMsg'
Learn More
Learn More
α-Methyl-D-mannopyranoside, 99+%, Thermo Scientific Chemicals
alpha-Methyl-D-mannopyranoside, 99+%, Acros Organics, Quantity: 500g, Packaging: Glass bottle, Gray or White, Melting Point: 186.0 deg.C to 194.0 deg.C, Molecular Weight: 194.18, Percent Purity: 99+%, Appearance: (1% aq. soln.) clear, colorless, Assay Percent Range: 99% min. (ELSD) (HPLC)
£28.40 - £273.00
Chemical Identifiers
CAS | 617-04-9 |
---|---|
Molecular Formula | C7H14O6 |
Molecular Weight (g/mol) | 194.18 |
MDL Number | MFCD00063262 |
InChI Key | HOVAGTYPODGVJG-UHFFFAOYNA-N |
Synonym | methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside |
PubChem CID | 101798 |
ChEBI | CHEBI:43943 |
IUPAC Name | (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
SMILES | COC1OC(CO)C(O)C(O)C1O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10229873
|
Thermo Scientific Acros
229250250 |
25 g | Glass bottle |
£28.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10023170
|
Thermo Scientific Acros
229251000 |
100 g | Glass bottle |
£77.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10720575
|
Thermo Scientific Acros
229255000 |
500 g | Glass bottle |
£273.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
617-04-9 | |
194.18 | |
HOVAGTYPODGVJG-UHFFFAOYNA-N | |
101798 | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol |
C7H14O6 | |
MFCD00063262 | |
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
CHEBI:43943 | |
COC1OC(CO)C(O)C(O)C1O |
Specifications
617-04-9 | |
100.0 | |
Gray or White | |
99% min. (ELSD) (HPLC) | |
MFCD00063262 | |
+ 79.00 | |
(10% in water) Clear colorless | |
COC1OC(CO)C(O)C(O)C1O | |
194.18 | |
CHEBI:43943 | |
99+% | |
α-Methyl-D-mannopyranoside |
99.0 | |
186.0°C to 194.0°C | |
Authentic | |
C7H14O6 | |
+ 79.00 (20.00°C c=10,H2O) | |
methyl a-d-mannopyranoside, methyl alpha-d-mannopyranoside, methyl alpha-d-mannoside, alpha-methyl-d-mannoside, alpha-methyl mannopyranoside, 1-o-methyl-alpha-d-mannopyranoside, methyl-alpha-d-mannoside, alpha-d-methyl mannoside, o1-methyl-mannose, alpha-methyl-d---mannoside | |
HOVAGTYPODGVJG-UHFFFAOYNA-N | |
(2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol | |
101798 | |
194.18 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 210-502-3
RUO – Research Use Only