missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl ethyl ketone, BAKER ANALYZED™ Laboratory Reagent, J.T. Baker™
102.60 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
1 L
Specifications
CAS | 78-93-3 |
Acidity | max. 0.003 % (as CH3COOH) |
Density | 0.797 to 801.000 g/mL at 25°C |
Boiling Point | 79.6°C |
Quantity | 1 L |
Assay Percent Range | min. 99 % |
Residue after Evaporation | max. 0.005 % |
Molecular Formula | C4H8O |
MDL Number | MFCD00011648 |
Water | max. 0.2 % |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'