missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Formate, 99%, Spectrophotometric Grade, Honeywell™ Riedel-de-Haën™
Click to view available options
Quantity:
1 L
Packaging:
Glass bottle
Description
- Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 107-31-3 |
Density | 0.974g/mL at 20°C |
Flash Point | −19°C (−2.2°F) |
Packaging | Glass bottle |
Molecular Formula | C2H4O2 |
Linear Formula | HCO2CH3 |
MDL Number | MFCD00003291 |
Quantity | 1 L |
UN Number | UN1243 |
Beilstein | 1734623 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'