missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl hexanoate, 99%, analytical standard for GC, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifica
CAS | 106-70-7 |
Melting Point | -71.0°C |
Density | 0.8840g/mL |
Boiling Point | 151.0°C |
Flash Point | 42°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (GC) |
Packaging | Glass bottle |
Molecular Formula | C7H14O2 |
Linear Formula | CH3(CH2)4COOCH3 |
Vedi altri risultati |
Sicurezza e movimentazione
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
Identificativo prodotto
- Methyl hexanoate
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'