Learn More
Methyl hexanoate, 99%, Thermo Scientific Chemicals
£28.30 - £113.00
Chemical Identifiers
CAS | 106-70-7 |
---|---|
Molecular Formula | C7H14O2 |
Molecular Weight (g/mol) | 130.19 |
MDL Number | MFCD00009510 |
InChI Key | NUKZAGXMHTUAFE-UHFFFAOYSA-N |
Synonym | methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate |
PubChem CID | 7824 |
ChEBI | CHEBI:77322 |
IUPAC Name | methyl hexanoate |
SMILES | CCCCCC(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499642
|
Thermo Scientific Alfa Aesar
A10757.18 |
50 g |
£28.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409652
|
Thermo Scientific Alfa Aesar
A10757.30 |
250 g |
£42.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489642
|
Thermo Scientific Alfa Aesar
A10757.0B |
1000 g |
£113.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-70-7 | |
130.19 | |
NUKZAGXMHTUAFE-UHFFFAOYSA-N | |
7824 | |
methyl hexanoate |
C7H14O2 | |
MFCD00009510 | |
methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate | |
CHEBI:77322 | |
CCCCCC(=O)OC |
Specifications
106-70-7 | |
0.885 | |
45°C (113°F) | |
1.405 | |
UN3272 | |
methyl caproate, hexanoic acid methyl ester, hexanoic acid, methyl ester, methyl hexoate, methyl capronate, methyl n-hexanoate, methyl hexylate, caproic acid methyl ester, methyl caproate natural, methyl n-hexoate | |
CCCCCC(=O)OC | |
130.19 | |
CHEBI:77322 | |
99% |
-71°C | |
150°C to 151°C | |
C7H14O2 | |
MFCD00009510 | |
1744683 | |
NUKZAGXMHTUAFE-UHFFFAOYSA-N | |
methyl hexanoate | |
7824 | |
130.19 | |
Methyl hexanoate |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
missing translation for 'einecsNumber' : 203-425-1
missing translation for 'rtecsNumber' : MO8401400
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only