missing translation for 'onlineSavingsMsg'
Learn More
Methyl hydrogen terephthalate, 99+%, Thermo Scientific Chemicals Product Code.: 15460698

Methyl hydrogen terephthalate, 99+%, Thermo Scientific Chemicals

Product Code. 15460698
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 15460698

Brand: Thermo Scientific Alfa Aesar H54391.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

Solubility
Slightly soluble in water. Also soluble in methanol, benzene and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-64-7
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00002557
InChI Key REIDAMBAPLIATC-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl benzoic acid, mono-methyl terephthalate, monomethyl terephthalate, methyl terephthalate, methyl hydrogen terephthalate, 1,4-benzenedicarboxylic acid, monomethyl ester, terephthalic acid monomethyl ester, 1,4-benzenedicarboxylic acid monomethyl ester, terephthalic acid, monomethyl ester, hydrogen methyl terephthalateShow MoreShow Less
PubChem CID 15513
IUPAC Name 4-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 220°C to 223°C
Quantity 100 g
Beilstein 1911082
Solubility Information Slightly soluble in water. Also soluble in methanol,benzene and ether.
Formula Weight 180.16
Percent Purity ≥99%
Chemical Name or Material Methyl hydrogen terephthalate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ15K009 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberH54391.06,H54391.22,H54391.14
Lot NumberR26I026 Certificate TypeCertificate of Analysis Date03/06/2022 Catalog NumberH54391.06,H54391.22,H54391.14
Lot Number81500374 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54391.06,H54391.22,H54391.14
Lot Number81400401 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54391.06,H54391.22,H54391.14
Lot Number81500222 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH54391.06,H54391.22,H54391.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl hydrogen terephthalate, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.