Learn More
Methyl indole-3-carboxylate, 99%, Thermo Scientific Chemicals
Description
Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reactant used for preparation of nitric oxide synthesis (nNOS) inhibitors, protein kinase c alpha (PKCα) inhibitors, inhibitors of the C-terminal domain of RNA polymerase II as antitumor agents, kinase insert domain receptor (KDR) inhibitors, organocatalysts for the anti-Mannich reaction, very late antigen-4 (VLA-4) antagonists, inhibitors of Human 5-Lipoxygenase, serotonin 5-HT4 receptor antagonists and hyaluronidase inhibitors.
Solubility
Slightly soluble methanol and dimethyl sulfoxide. Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 149°C to 152°C |
Quantity | 25 g |
Beilstein | 142023 |
Solubility Information | Slightly soluble methanol and dimethyl sulfoxide. Insoluble in water. |
Formula Weight | 175.19 |
Percent Purity | 99% |
Chemical Name or Material | Methyl indole-3-carboxylate |
RUO – Research Use Only