missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl indole-5-carboxylate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin?s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin′s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Reactant for biosynthesis of inhibitors of protein kinases, reactant in metal-free Friedel-Crafts alkylation, reactant in preparation of diphenylsulfonium ylides from Martin′s sulfurane, reactant in cross dehydrogenative coupling reactions, reactant in synthesis of indirubin derivatives, reactant in preparation of aminoindolylacetates.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 123°C to 126°C |
Quantity | 5 g |
Beilstein | 474256 |
Solubility Information | Insoluble in water. |
Formula Weight | 175.19 |
Percent Purity | ≥98% |
Chemical Name or Material | Methyl indole-5-carboxylate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'