missing translation for 'onlineSavingsMsg'
Learn More

Methyl isobutyrate, 98%, Thermo Scientific Chemicals

£20.90 - £234.00

Chemical Identifiers

CAS 547-63-7
Molecular Formula C5H10O2
Molecular Weight (g/mol) 102.133
MDL Number MFCD00008914
InChI Key BHIWKHZACMWKOJ-UHFFFAOYSA-N
Synonym methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
PubChem CID 11039
ChEBI CHEBI:73689
IUPAC Name methyl 2-methylpropanoate
SMILES CC(C)C(=O)OC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11477996
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A14070.AE
100 mL
£20.90
100mL
Please to purchase this item. Need a web account? Register with us today!
11487996
View Documents
Thermo Scientific Alfa Aesar
A14070.AP
500 mL
£74.30
500mL
Please to purchase this item. Need a web account? Register with us today!
11497996
View Documents
Thermo Scientific Alfa Aesar
A14070.0F
2500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Methyl isobutyrate is used as a flavoring agent. It was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl isobutyrate is used as a flavoring agent. It was used as diluent in forced Rayleigh scattering studies to measure diffusion coefficients for a ternary system comprising camphorquinone in poly(methyl methacrylate).

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

547-63-7
102.133
BHIWKHZACMWKOJ-UHFFFAOYSA-N
11039
methyl 2-methylpropanoate
C5H10O2
MFCD00008914
methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
CHEBI:73689
CC(C)C(=O)OC

Specifications

547-63-7
0.891
3°C (37°F)
C5H10O2
MFCD00008914
1740720
methyl isobutyrate, methyl 2-methylpropionate, methyl isobutanoate, propanoic acid, 2-methyl-, methyl ester, isobutyric acid, methyl ester, methylisobutyrate, isobutyric acid methyl ester, poly d-lactide, methyl isobutyrate natural, methylester kyseliny isomaselne
BHIWKHZACMWKOJ-UHFFFAOYSA-N
methyl 2-methylpropanoate
11039
102.13
Methyl isobutyrate
-85°C
90°C to 92°C
Pleasant
1.384
UN1237
14,6088
Slightly soluble in water.
CC(C)C(=O)OC
102.133
CHEBI:73689
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H332
Highly flammable liquid and vapour.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYL BUTYRATE

missing translation for 'einecsNumber' : 208-929-5

missing translation for 'rtecsNumber' : NQ5425000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only