missing translation for 'onlineSavingsMsg'
Learn More

Methyl levulinate, 99+%, Thermo Scientific Chemicals

Product Code. 11315677
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl levulinate is mainly used as raw material on perfume.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-45-3
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00017499
InChI Key UAGJVSRUFNSIHR-UHFFFAOYSA-N
Synonym methyl levulinate, methyllevulinate, levulinic acid methyl ester, pentanoic acid, 4-oxo-, methyl ester, methyl 4-oxovalerate, levulinic acid, methyl ester, 4-oxovaleric acid methyl ester, 4-oxo-pentanoic acid methyl ester, methyl levulate, me ester
PubChem CID 69354
IUPAC Name methyl 4-oxopentanoate
SMILES CC(=O)CCC(=O)OC

Specifications

Density 1.05 g/mL
Boiling Point 191°C
Flash Point 72°C (161.6°F)
Odor Pleasant
Quantity 500 g
Solubility Information It is soluble in ethanol,ether,acetone and benzene.
Formula Weight 130.14
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Methyl levulinate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'