missing translation for 'onlineSavingsMsg'
Learn More

Methyl linolenate, 99%

Product Code. 10079001 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10079001

Brand: Thermo Scientific Acros 368100050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 301-00-8 | C19H32O2 | 292.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 301-00-8
Molecular Formula C19H32O2
Molecular Weight (g/mol) 292.46
MDL Number MFCD00135851
InChI Key DVWSXZIHSUZZKJ-YSTUJMKBSA-N
Synonym methyl linolenate, linolenic acid methyl ester, methyl alpha-linolenate, linolenic acid, methyl ester, unii-0s1ns923k6, alpha-linolenic acid methyl ester, methyl all-cis-9,12,15-octadecatrienoate, methyl 9z,12z,15z-octadeca-9,12,15-trienoate, 9,12,15-octadecatrienoic acid, methyl ester, z,z,z, methyl cis,cis,cis-octadec-9,12,15-trienoate
PubChem CID 5319706
IUPAC Name methyl (9Z,12Z,15Z)-octadeca-9,12,15-trienoate
SMILES CC\C=C/C\C=C/C\C=C/CCCCCCCC(=O)OC

Specifications

Color Colorless
Density 0.8950g/mL
Boiling Point 182.0°C (3.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2CH=CH)3(CH2)7CO2CH3
Refractive Index 1.4700 to 1.4720
Quantity 5 g
Beilstein 02, IV, 1782
Specific Gravity 0.895
Solubility Information Solubility in water: insoluble
Formula Weight 292.46
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Methyl linolenate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.