missing translation for 'onlineSavingsMsg'
Learn More

Methyl malonyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11400510
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl malonyl chloride is used as pharmaceutical intermediate. Used in anti-cancer drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37517-81-0
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.531
MDL Number MFCD00013657
InChI Key UTBCRHAMJFMIIR-UHFFFAOYSA-N
PubChem CID 123460
IUPAC Name methyl 3-chloro-3-oxopropanoate
SMILES COC(=O)CC(=O)Cl

Specifications

Color Colorless
Density 1.27 g/mL
Boiling Point 58°C (12 mmHg)
Flash Point 80°C (176°F)
Refractive Index 1.4325
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym methyl malonyl chloride, methyl 3-chloro-3-oxopropionate, methyl chloroformylacetate, propanoic acid, 3-chloro-3-oxo-, methyl ester, methyl-3-chloro-3-oxopropionate, methyl-3-chlor-3-oxopropanoat, methyl chlorocarbonylacetate, carbomethoxy acetyl chloride, methyl chloroformyl acetate, methyl3-chloro-3-oxopropionate
Solubility Information Reacts with water.
Formula Weight 136.53
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Methyl malonyl chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'