Learn More
Methyl mercaptoacetate, 95%, Thermo Scientific Chemicals
£20.80 - £58.10
Chemical Identifiers
CAS | 2365-48-2 |
---|---|
Molecular Formula | C3H6O2S |
Molecular Weight (g/mol) | 106.14 |
MDL Number | MFCD00004873 |
InChI Key | MKIJJIMOAABWGF-UHFFFAOYSA-N |
Synonym | methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el |
PubChem CID | 16907 |
IUPAC Name | methyl 2-sulfanylacetate |
SMILES | COC(=O)CS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11366646
|
Thermo Scientific Alfa Aesar
L03783.14 |
25 g |
£20.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11376646
|
Thermo Scientific Alfa Aesar
L03783.22 |
100 g |
£23.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11386646
|
Thermo Scientific Alfa Aesar
L03783.36 |
500 g |
£58.10
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl thioglycolate was used in the preparation of 3-carbomethoxy-4- oxotetrahydrothiopyran, 2- and 4-carbomethoxy-3-oxotetrahydrothiophene. It is used to prepare methyl thioglycolate and aminoethanethiol conjugated gold nanorods.
Solubility
Soluble in water. (40 g/L at 20°C)
Notes
Air Sensitive. Store under dry inert gas. Incompatible with oxidizing agents and air.
Chemical Identifiers
2365-48-2 | |
106.14 | |
MKIJJIMOAABWGF-UHFFFAOYSA-N | |
16907 | |
COC(=O)CS |
C3H6O2S | |
MFCD00004873 | |
methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el | |
methyl 2-sulfanylacetate |
Specifications
2365-48-2 | |
148°C to 149°C | |
Stench | |
HSCH2COOCH3 | |
UN1228 | |
methyl thioglycolate, methyl mercaptoacetate, methyl 2-mercaptoacetate, methylthioglycolate, thioglycolic acid methyl ester, acetic acid, mercapto-, methyl ester, methanethiol, methoxy, usaf ek-7119, thioglykolsaeure-methylester, unii-o1608la9el | |
COC(=O)CS | |
106.14 | |
106.14 | |
Methyl mercaptoacetate |
-3°C | |
56°C (132°F) | |
C3H6O2S | |
MFCD00004873 | |
506259 | |
MKIJJIMOAABWGF-UHFFFAOYSA-N | |
methyl 2-sulfanylacetate | |
16907 | |
98% |
Safety and Handling
GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.
missing translation for 'rtecsNumber' : AI7350000
missing translation for 'tsca' : TSCA
RUO – Research Use Only