missing translation for 'onlineSavingsMsg'
Learn More

Methyl methacrylate, 99%, stab.

Product Code. 11499953
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
This item is not returnable. View return policy

Product Code. 11499953

Brand: Thermo Scientific Alfa Aesar A13030.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80-62-6 | C5H8O2 | 100.117 g/mol

Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl methacrylate is used as a monomer in the manufacturing of poly(methyl methacrylate) (PMMA). It is also used in the manufacturing of the co-polymer methyl methacrylate-butadiene-styrene (MBS), which is a modifier for polyvinyl chloride. It plays an important role as the cement used in total hip replacements as well as total knee replacements. It is also involved in the preparation of other methacrylates viz. ethyl methacrylate (EMA), butyl methacrylate (BMA) and 2-ethyl hexyl methacrylate (2-EHMA) and methacrylic acid (MAA), which finds application as an intermediate in the manufacture of coating polymers and textile applications.

Solubility
Slightly miscible with water.

Notes
Incompatible with oxidizing agents, peroxides, amines, bases, acids, reducing agents and halogens.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-62-6
Molecular Formula C5H8O2
MDL Number MFCD00008587
InChI Key VVQNEPGJFQJSBK-UHFFFAOYSA-N
Synonym methyl methacrylate, methylmethacrylate, methacrylic acid methyl ester, methyl methylacrylate, methyl 2-methylpropenoate, pegalan, methyl-methacrylat, diakon, acryester m, methyl 2-methyl-2-propenoate
PubChem CID 6658
ChEBI CHEBI:34840
IUPAC Name methyl 2-methylprop-2-enoate

Specifications

Melting Point -48°C
Density 0.939
Boiling Point 100°C to 101°C
Flash Point 10°C (50°F)
Refractive Index 1.415
Quantity 500 mL
UN Number UN1247
Beilstein 605459
Merck Index 14,5941
Solubility Information Slightly miscible with water.
SMILES CC(=C)C(=O)OC
Molecular Weight (g/mol) 100.117
Formula Weight 100.12
Percent Purity 99%
Chemical Name or Material Methyl methacrylate, stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Methyl methacrylate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methyl methacrylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.