missing translation for 'onlineSavingsMsg'
Learn More

Methyl methylphenylphosphinate, 98%, Thermo Scientific Chemicals

Product Code. 11413038
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6389-79-3
Molecular Formula C8H11O2P
Molecular Weight (g/mol) 170.15
MDL Number MFCD00013909
InChI Key RTAAKNLNWIHGGX-UHFFFAOYNA-N
Synonym methyl methylphenylphosphinate, methyl methyl phenyl phosphinate, methoxy methyl phosphoryl benzene, acmc-1b8qc, methylphenylphosphinic acid methyl ester, phenyl methyl phosphinic acid methyl ester, +-methylphenylphosphinic acid methyl ester, ?-methylphenylphosphinic acid methyl ester, phosphinic acid, methylphenyl-, methyl ester, r
PubChem CID 323835
IUPAC Name [methoxy(methyl)phosphoryl]benzene
SMILES COP(C)(=O)C1=CC=CC=C1

Specifications

Boiling Point 148°C to 149°C (17 mmHg)
Quantity 5 g
Formula Weight 170.15
Percent Purity 98%
Chemical Name or Material Methyl methylphenylphosphinate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.