missing translation for 'onlineSavingsMsg'
Learn More
Learn More
methyl N-(3,5-dichlorophenyl)carbamate, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 25 g |
Formula Weight | 220.05 |
Percent Purity | 97% |
Chemical Name or Material | Methyl N-(3, 5-dichlorophenyl)carbamate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'