missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl N-methylanthranilate, 98%, Thermo Scientific Chemicals
£36.70 - £111.00
Chemical Identifiers
CAS | 85-91-6 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
MDL Number | MFCD00017183 |
InChI Key | GVOWHGSUZUUUDR-UHFFFAOYSA-N |
Synonym | methyl 2-methylamino benzoate, methyl n-methylanthranilate, dimethyl anthranilate, methyl methylaminobenzoate, methyl methanthranilate, methyl methylanthranilate, benzoic acid, 2-methylamino-, methyl ester, methyl n-methyl anthranilate, methyl n-methyl-o-anthranilate, methyl o-methylamino benzoate |
PubChem CID | 6826 |
IUPAC Name | methyl 2-(methylamino)benzoate |
SMILES | CNC1=CC=CC=C1C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11409706
|
Thermo Scientific Alfa Aesar
B20233.18 |
50 g |
£36.70
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419706
|
Thermo Scientific Alfa Aesar
B20233.30 |
250 g |
£111.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl N-methylanthranilate is a fragrance ingredient used in decorative cosmetics, fine fragrances, shampoos, toilet soaps and other toiletries as well as in non-cosmetic products such as household cleaners and detergents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl N-methylanthranilate is a fragrance ingredient used in decorative cosmetics, fine fragrances, shampoos, toilet soaps and other toiletries as well as in non-cosmetic products such as household cleaners and detergents.
Solubility
Miscible in all proportions with ethanol 96%, DMSO and diethyl ether.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
85-91-6 | |
165.19 | |
GVOWHGSUZUUUDR-UHFFFAOYSA-N | |
6826 | |
CNC1=CC=CC=C1C(=O)OC |
C9H11NO2 | |
MFCD00017183 | |
methyl 2-methylamino benzoate, methyl n-methylanthranilate, dimethyl anthranilate, methyl methylaminobenzoate, methyl methanthranilate, methyl methylanthranilate, benzoic acid, 2-methylamino-, methyl ester, methyl n-methyl anthranilate, methyl n-methyl-o-anthranilate, methyl o-methylamino benzoate | |
methyl 2-(methylamino)benzoate |
Specifications
85-91-6 | |
256°C to 258°C | |
C9H11NO2 | |
607217 | |
GVOWHGSUZUUUDR-UHFFFAOYSA-N | |
methyl 2-(methylamino)benzoate | |
6826 | |
98% |
17°C to 19°C | |
133°C (271°F) | |
MFCD00017183 | |
methyl 2-methylamino benzoate, methyl n-methylanthranilate, dimethyl anthranilate, methyl methylaminobenzoate, methyl methanthranilate, methyl methylanthranilate, benzoic acid, 2-methylamino-, methyl ester, methyl n-methyl anthranilate, methyl n-methyl-o-anthranilate, methyl o-methylamino benzoate | |
CNC1=CC=CC=C1C(=O)OC | |
165.19 | |
165.19 | |
Methyl N-methylanthranilate |
Safety and Handling
missing translation for 'rtecsNumber' : CB3500000
missing translation for 'tsca' : TSCA
RUO – Research Use Only