Learn More
Methyl oleate, 96%, Thermo Scientific Chemicals
£73.87 - £194.97
Chemical Identifiers
CAS | 112-62-9 |
---|---|
Molecular Formula | C19H36O2 |
Molecular Weight (g/mol) | 296.495 |
MDL Number | MFCD00009578 |
InChI Key | QYDYPVFESGNLHU-KHPPLWFESA-N |
Synonym | methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 |
PubChem CID | 5364509 |
ChEBI | CHEBI:27542 |
IUPAC Name | methyl (Z)-octadec-9-enoate |
SMILES | CCCCCCCCC=CCCCCCCCC(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15434077
|
Thermo Scientific Alfa Aesar
H31358.14 |
25 g |
£73.87
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15444077
|
Thermo Scientific Alfa Aesar
H31358.22 |
100 g |
£194.97
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Methyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers for duplicating inks, rubbers and waxes. It finds application as lubricants and lubricant additives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsMethyl oleate is used as a chromatographic reference standard in biochemical research. It is also used as an intermediate for detergents, emulsifiers, wetting agents, stabilizers, textile treatments, plasticizers for duplicating inks, rubbers and waxes. It finds application as lubricants and lubricant additives.
Solubility
Miscible with alcohol, ether and chloroform. Immiscible with water.
Notes
Air and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
Chemical Identifiers
112-62-9 | |
296.495 | |
QYDYPVFESGNLHU-KHPPLWFESA-N | |
5364509 | |
methyl (Z)-octadec-9-enoate |
C19H36O2 | |
MFCD00009578 | |
methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 | |
CHEBI:27542 | |
CCCCCCCCC=CCCCCCCCC(=O)OC |
Specifications
112-62-9 | |
168°C to 170°C (2 mmHg) | |
C19H36O2 | |
1.452 | |
1727037 | |
methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 | |
QYDYPVFESGNLHU-KHPPLWFESA-N | |
methyl (Z)-octadec-9-enoate | |
5364509 | |
296.5 | |
Methyl oleate |
0.876 | |
163°C (325°F) | |
CH3(CH2)7CH=CH(CH2)7CO2CH3 | |
MFCD00009578 | |
14,6828 | |
Miscible with alcohol,ether and chloroform. Immiscible with water. | |
CCCCCCCCC=CCCCCCCCC(=O)OC | |
296.495 | |
CHEBI:27542 | |
96% |
Safety and Handling
missing translation for 'einecsNumber' : 203-992-5
missing translation for 'rtecsNumber' : RK0895000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.