missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl oleate, technical, Thermo Scientific Chemicals
£22.20 - £129.00
Chemical Identifiers
CAS | 67762-38-3 |
---|---|
Molecular Formula | C19H36O2 |
Molecular Weight (g/mol) | 296.50 |
MDL Number | MFCD00009578 |
InChI Key | QYDYPVFESGNLHU-ZHACJKMWSA-N |
Synonym | methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 |
PubChem CID | 5364509 |
ChEBI | CHEBI:27542 |
IUPAC Name | methyl (Z)-octadec-9-enoate |
SMILES | CCCCCCCCC=CCCCCCCCC(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10760672
|
Thermo Scientific Acros
414645000 |
500 g | Glass bottle |
£22.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10760482
|
Thermo Scientific Acros
414640010 |
1 kg | Glass bottle |
£36.40
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10434315
|
Thermo Scientific Acros
414640050 |
5 kg | Plastic drum |
£129.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
67762-38-3 | |
296.50 | |
QYDYPVFESGNLHU-ZHACJKMWSA-N | |
5364509 | |
methyl (Z)-octadec-9-enoate |
C19H36O2 | |
MFCD00009578 | |
methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 | |
CHEBI:27542 | |
CCCCCCCCC=CCCCCCCCC(=O)OC |
Specifications
67762-38-3 | |
Yellow to Amber | |
186.0°C (9.0 mmHg) | |
Authentic | |
CH3(CH2)7CHCH(CH2)7COOCH3 | |
MFCD00009578 | |
methyl oleate, oleic acid methyl ester, methyl cis-9-octadecenoate, oleic acid, methyl ester, edenor metio5, methyl z-9-octadecenoate, methyl-cis-oleate, exceparl m-ol, emery, oleic acid ester, esterol 112 | |
QYDYPVFESGNLHU-ZHACJKMWSA-N | |
methyl (Z)-octadec-9-enoate | |
5364509 | |
6.829 mPa.s (20°C) | |
≥55% | |
Oily Liquid |
-20.0°C | |
0.8700g/mL | |
180°C | |
C19H36O2 | |
1.4510 to 1.4580 | |
0.87 | |
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents | |
CCCCCCCCC=CCCCCCCCC(=O)OC | |
296.50 | |
CHEBI:27542 | |
296.48 | |
Technical | |
Methyl oleate |
Safety and Handling
missing translation for 'einecsNumber' : 203-992-5
Recommended Storage : Refrigerator +4°C
RUO – Research Use Only