missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Methyl Orange Product Code.: 11498227

Thermo Scientific Chemicals Methyl Orange

Product Code. 11498227
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11498227

Brand: Thermo Scientific Chemicals A17604.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl Orange is a pH indicator used in titrations for acids. It is also used for histological microscopy. Further, it is used as counter stain to crystal violet in staining pollen tubes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl Orange is a pH indicator used in titrations for acids. It is also used for histological microscopy. Further, it is used as counter stain to crystal violet in staining pollen tubes.

Solubility
Soluble in ethanol. Partially soluble in hot water. Slightly soluble in cold water and pyrimidine. Insoluble in ether and alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 547-58-0
Molecular Formula C14H14N3NaO3S
Molecular Weight (g/mol) 327.334
MDL Number MFCD00007502
InChI Key STZCRXQWRGQSJD-UHFFFAOYSA-M
Synonym Acid Orange 52; C.I. 13025
PubChem CID 23673835
IUPAC Name sodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Melting Point >300°C
UN Number UN3143
Beilstein 4732884
Merck Index 14,6105
Solubility Information Soluble in ethanol. Partially soluble in hot water. Slightly soluble in cold water and pyrimidine. Insoluble in ether and alcohol.
Formula Weight 327.34
Odor Weak, Characteristic
pH 6.5
Quantity 500 g
Chemical Name or Material Methyl Orange

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251782 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA17604.22,A17604.36
Lot Number10249345 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog NumberA17604.22,A17604.36
Lot Number10243233 Certificate TypeCertificate of Analysis Date09/09/2022 Catalog NumberA17604.22,A17604.36
Lot Number10237639 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA17604.22,A17604.36
Lot Number10162983 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17604.22,A17604.36
compliance-icons
Product Identifier
  • Methyl Orange, ACS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Supplemental information
  • MIXTURE LIST-Contains : C.I. Acid Orange 52

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Methyl Orange >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.