missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Indicator 0.1% Alcoholic Solution
High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
Brand: Reagecon MTR05025
This item is not returnable.
View return policy
Description
- Titrant, Standard Titrant, Standard Solution, Standardized Solution, Reference Standard, Reference Material
- For manual titrations; indicator titrant or analyte change color and this color change are by far the most important method of end point detection.
- This high-quality indicator solution facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Accuracy ± 0.1% | |
HDPE Flip-Top Bottle | |
Titration and Analyte Color Change |
0.001 | |
250 mL | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- INDICATOR METHYL ORANGE
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : ethyl alcohol, methyl orange