missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange Indicator, Chem Lab™
Specifications
Melting Point | 0°C |
---|---|
Solubility Information | Soluble in water |
Odor | Odorless |
Color | Orange |
Physical Form | Liquid |
Specifications
0°C | |
Odorless | |
Liquid |
Soluble in water | |
Orange |