missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl Orange solution, Honeywell Fluka™
Click to view available options
Quantity:
500 mL
1 L
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
UN Number | UN1993 |
Beilstein | 4732884 |
Formula Weight | 327.33g/mol |
Percent Purity | 0.10% |
Quantity | 500 mL |
Flash Point | 39°C (102.2°F) |
Chemical Name or Material | Methyl Orange solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'