missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Orange-xylene cyanol solution in water, pure
Click to view available options
Quantity:
500 mL
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 62758-15-0 |
Molecular Formula | C39H41N5Na2O10S3 |
MDL Number | MFCD00077269 |
InChI Key | DLSJBPNVZNTATN-MXBKRXPOSA-L |
SMILES | CCNC1=C(C=C(C=C1)C(=C2C=CC(=NCC)C(=C2)C)C3=CC(=C(C=C3S(=O)(=O)O)S(=O)(=O)[O-])O)C.CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+].[Na+] |
IUPAC Name | disodium;4-[[4-(dimethylamino)phenyl]diazenyl]benzenesulfonate;4-[(E)-[4-(ethylamino)-3-methylphenyl]-(4-ethylimino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-hydroxy-5-sulfobenzenesulfonate |
Molecular Weight (g/mol) | 881.942 |
PubChem CID | 90474202 |
Formula Weight | 865.96 |
Grade | Pure |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'