Learn More
Methyl orotate, 98%, ACROS Organics™
£177.00 - £668.00
Chemical Identifiers
CAS | 6153-44-2 |
---|---|
Molecular Formula | C6H6N2O4 |
Molecular Weight (g/mol) | 170.12 |
MDL Number | MFCD00010564 |
InChI Key | UUTDWTOZAWFKFW-UHFFFAOYSA-N |
Synonym | methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate |
PubChem CID | 80257 |
IUPAC Name | methyl 2,4-dioxo-1H-pyrimidine-6-carboxylate |
SMILES | COC(=O)C1=CC(=O)NC(=O)N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10104840
|
Thermo Scientific Acros
208780250 |
25 g | Glass bottle |
£177.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10444145
|
Thermo Scientific Acros
208781000 |
100 g | Glass bottle |
£668.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6153-44-2 | |
170.12 | |
UUTDWTOZAWFKFW-UHFFFAOYSA-N | |
80257 | |
COC(=O)C1=CC(=O)NC(=O)N1 |
C6H6N2O4 | |
MFCD00010564 | |
methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate | |
methyl 2,4-dioxo-1H-pyrimidine-6-carboxylate |
Specifications
6153-44-2 | |
Brown or White to Yellow | |
95% | |
MFCD00010564 | |
15, 6972 | |
Solubility in water: .. Other solubilities: soluble in 1n nh4oh | |
COC(=O)C1=CC(=O)NC(=O)N1 | |
170.12 | |
170.12 | |
Powder |
244°C | |
Authentic | |
C6H6N2O4 | |
25, 254 | |
methyl orotate, orotic acid methyl ester, methyl 2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate, unii-41c6ifg4mq, 4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-2,6-dioxo-, methyl ester, 6-carbomethyoxyuracil, 41c6ifg4mq, methyl 2,6-dihydroxypyrimidine-4-carboxylate, methyl 2,6-dihydroxy-4-pyrimidinecarboxylate, methyl 2,6-dioxo-1,3-dihydropyrimidine-4-carboxylate | |
UUTDWTOZAWFKFW-UHFFFAOYSA-N | |
methyl 2,4-dioxo-1H-pyrimidine-6-carboxylate | |
80257 | |
95% | |
Methyl orotate |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lens
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 228-171-9
RUO – Research Use Only