missing translation for 'onlineSavingsMsg'
Learn More
Methyl-p-benzoquinone, 98%, Thermo Scientific Chemicals Product Code.: 11344187

Methyl-p-benzoquinone, 98%, Thermo Scientific Chemicals

Product Code. 11344187
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11344187

Brand: Thermo Scientific Alfa Aesar 036670.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methyl-p-benzoquinone imine is the products were the tetrahedral adducts. Synthesis of 2H-indazole-4,7-dione derivatives from3-phenylsydnone and p-toluquinone. A Michael addition of 2-methylcyclopentane-1,3-dione to various p-toluquinone imides exclusively afforded the sole products in a regiospecific manner.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-97-9
Molecular Formula C7H6O2
Molecular Weight (g/mol) 122.12
MDL Number MFCD00001603
InChI Key VTWDKFNVVLAELH-UHFFFAOYSA-N
Synonym p-toluquinone, methyl-p-benzoquinone, 2-methyl-1,4-benzoquinone, toluquinone, methyl-1,4-benzoquinone, 2-methyl-p-benzoquinone, tolylquinone, 2-methylbenzoquinone, 2-methylquinone, methylbenzoquinoneShow MoreShow Less
PubChem CID 11122
IUPAC Name 2-methylcyclohexa-2,5-diene-1,4-dione
SMILES CC1=CC(=O)C=CC1=O

Specifications

Melting Point 67°C to 70°C
Color Green-Yellow
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 122.12
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material Methyl-p-benzoquinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU27K052 Certificate TypeCertificate of Analysis Date11/09/2024 Catalog Number036670.18,036670.09
Lot NumberT09K026 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog Number036670.18,036670.09
Lot NumberZ11J002 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog Number036670.18,036670.09
Lot NumberM25I027 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number036670.18,036670.09
Lot NumberG31S059 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number036670.18,036670.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Methyl-p-benzoquinone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.