missing translation for 'onlineSavingsMsg'
Learn More

Methyl piperidine-4-carboxylate, 98%, Thermo Scientific Chemicals

Product Code. 15408328
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2971-79-1
Molecular Formula C7H13NO2
Molecular Weight (g/mol) 143.19
MDL Number MFCD00190578
InChI Key RZVWBASHHLFBJF-UHFFFAOYSA-N
Synonym methyl isonipecotate, methyl 4-piperidinecarboxylate, 4-piperidinecarboxylic acid, methyl ester, piperidine-4-carboxylic acid methyl ester, 4-piperidinecarboxylic acid methyl ester, isonipecotic acid methyl ester, methylpiperidine-4-carboxylate, methylisonipecotate, methyl piperidin-4-carboxylate, acmc-20aiv4
PubChem CID 424914
IUPAC Name methyl piperidine-4-carboxylate
SMILES COC(=O)C1CCNCC1

Specifications

Color Colorless to Yellow
Density 1.06 g/mL
Boiling Point 85°C to 90°C
Flash Point 89°C (192°F)
Refractive Index 1.465
Quantity 100 g
Beilstein 224128
Solubility Information Slightly soluble in water.
Formula Weight 143.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl piperidine-4-carboxylate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.