missing translation for 'onlineSavingsMsg'
Learn More

Methyl pyrrole-3-carboxylate, 97%, Thermo Scientific Chemicals

Product Code. 15400668
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl pyrrole-3-carboxylate is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2703-17-5
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00967074
InChI Key WLBNVSIQCFHAQB-UHFFFAOYSA-N
Synonym methyl pyrrole-3-carboxylate, 1h-pyrrole-3-carboxylic acid methyl ester, 1h-pyrrole-3-carboxylic acid, methyl ester, methyl 3-pyrrolecarboxylate, 3-methoxycarbonyl-1h-pyrrole, pubchem22507, acmc-1cmqn, methylpyrrole-3-carboxylate, 4,5-methoxy-carbonylpyrrole, methyl pyrrole 3-carboxylate
PubChem CID 12361759
IUPAC Name methyl 1H-pyrrole-3-carboxylate
SMILES COC(=O)C1=CNC=C1

Specifications

Melting Point 85°C to 89°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 125.13
Percent Purity 97%
Chemical Name or Material Methyl pyrrole-3-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'