missing translation for 'onlineSavingsMsg'
Learn More

Methyl (R)-(-)-β-hydroxyisobutyrate, 98%, Thermo Scientific Chemicals

Product Code. 10351630
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72657-23-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00063450
InChI Key ATCCIZURPPEVIZ-SCSAIBSYSA-N
Synonym r-methyl 3-hydroxy-2-methylpropanoate, r---3-hydroxy-2-methylpropionic acid methyl ester, methyl r---3-hydroxy-2-methylpropionate, --methyl d-beta-hydroxyisobutyrate, methyl r---3-hydroxyisobutyrate, roche ester, methyl r-3-hydroxy-2-methylpropionate, propanoic acid, 3-hydroxy-2-methyl-, methyl ester, 2r, methyl 2r-3-hydroxy-2-methylpropanoate, r---3-hydroxyisobutyric acid methyl ester
PubChem CID 5324733
IUPAC Name methyl (2R)-3-hydroxy-2-methylpropanoate
SMILES CC(CO)C(=O)OC

Specifications

Color Colorless
Density 1.0700g/mL
Boiling Point 60.0°C (4.0 Torr)
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH(CH3)COOCH3
Refractive Index 1.4243
Quantity 5 g
Specific Gravity 1.07
Specific Rotation − 23.30
Solubility Information Solubility in water: insoluble. Other solubilities: in ethanol: soluble
Formula Weight 118.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Methyl (R)-(-)-β-hydroxyisobutyrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'