Learn More
Description
Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Bacterial strains isolated from dye-contaminated sludge decolorized a toxic azo dye, namely, methyl red (MR). Methyl red is used in the methyl red test (MR test), used to identify bacteria producing stable acids by mechanisms of mixed acid fermentation of glucose
Solubility
Insoluble in water. Soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 179°C to 182°C |
Beilstein | 750102 |
Merck Index | 14,6119 |
Solubility Information | Insoluble in water. Soluble in ethanol. |
Formula Weight | 269.3 |
Quantity | 100 g |
Chemical Name or Material | Methyl Red |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253166 | Certificate of Analysis | 26/11/2024 | A16690.14,A16690.36,A16690.22 |
10251188 | Certificate of Analysis | 30/08/2024 | A16690.14,A16690.36,A16690.22 |
10247074 | Certificate of Analysis | 21/06/2023 | A16690,A16690.14,A16690.36,A16690.22 |
10242339 | Certificate of Analysis | 28/07/2022 | A16690.14,A16690.36,A16690.22 |
10239140 | Certificate of Analysis | 12/04/2022 | A16690.14,A16690.36,A16690.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.