missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl red, For ACS analysis, MP Biomedicals™
Click to view available options
Quantity:
250 g
Specifications
Formula Weight | 269.3 |
Color | Red |
Quantity | 250 g |
Physical Form | Powder |
Chemical Name or Material | Methyl red, For ACS analysis |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'