missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Methyl Red sodium salt, 0.02% w/v aq. soln.
£18.99 - £38.19
Chemical Identifiers
CAS | 845-10-3 |
---|---|
Molecular Formula | C15H14N3NaO2 |
Molecular Weight (g/mol) | 291.286 |
MDL Number | MFCD00002426 |
InChI Key | GNTPCYMJCJNRQB-UHFFFAOYSA-M |
PubChem CID | 4465632 |
IUPAC Name | sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
SMILES | CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11319668
|
Thermo Scientific Chemicals
044161.AE |
100 mL |
£18.99
100mL |
In Stock
Log in to see stock. |
|||||
11410690
|
Thermo Scientific Chemicals
044161.AP |
500 mL |
£38.19
500mL |
In Stock
Log in to see stock. |
|||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
845-10-3 | |
291.286 | |
GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate |
C15H14N3NaO2 | |
MFCD00002426 | |
4465632 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] |
Spezifikation
845-10-3 | |
C15H14N3NaO2 | |
GNTPCYMJCJNRQB-UHFFFAOYSA-M | |
sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate | |
4465632 | |
Odorless | |
Liquid |
0.02% w/v | |
MFCD00002426 | |
CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+] | |
291.286 | |
291.29 | |
Colorless | |
Methyl Red sodium salt |
Sicherheit und Handhabung
missing translation for 'einecsNumber' : 212-682-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only