missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Methyl Red sodium salt, ACS

Product Code. 11394177
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Methyl Red sodium salt is used as a pH indicator. In microbiology, it is used to make agar plates in bacteriology. Further, it is used as a coloring agent and also finds use in mouthwash, toothpaste and in cosmetics. It is also employed as a rocket propellant. In addition to this, it is used in the preparation of reversible stain for rapid detection of stained cells in each specimen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845-10-3
Molecular Formula C15H14N3NaO2
Molecular Weight (g/mol) 291.286
MDL Number MFCD00002426
InChI Key GNTPCYMJCJNRQB-UHFFFAOYSA-M
Synonym 2-[4-(Dimethylamino)phenyl]azobenzoic acid sodium salt
PubChem CID 4465632
IUPAC Name sodium;2-[[4-(dimethylamino)phenyl]diazenyl]benzoate
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=CC=C2C(=O)[O-].[Na+]

Specifications

Beilstein 4065488
Solubility Information Soluble in water. Slightly soluble in ethanol.
Formula Weight 291.29
Grade ACS
Color Brown to Red
Quantity 100 g
Physical Form Crystalline
Chemical Name or Material Methyl Red sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'