Learn More
Methyl sulfide, 99+%, extra pure, Thermo Scientific Chemicals
£23.62 - £89.04
Chemical Identifiers
CAS | 75-18-3 |
---|---|
Molecular Formula | C2H6S |
Molecular Weight (g/mol) | 62.13 |
MDL Number | MFCD00008562 |
InChI Key | QMMFVYPAHWMCMS-UHFFFAOYSA-N |
Synonym | dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane |
PubChem CID | 1068 |
ChEBI | CHEBI:17437 |
IUPAC Name | methylsulfanylmethane |
SMILES | CSC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10071470
|
Thermo Scientific Acros
127760100 |
10 mL | Glass bottle |
£23.62
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10071460
|
Thermo Scientific Acros
127760010 |
1 L | Glass bottle |
£49.45
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10264790
|
Thermo Scientific Acros
127760025 |
2.5 L | Glass bottle |
£89.04
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-18-3 | |
62.13 | |
QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
1068 | |
methylsulfanylmethane |
C2H6S | |
MFCD00008562 | |
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
CHEBI:17437 | |
CSC |
Specifications
75-18-3 | |
Colorless | |
38.0°C | |
Authentic | |
C2H6S | |
1.4351 | |
01, 288 | |
15, 3281 | |
dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethane | |
QMMFVYPAHWMCMS-UHFFFAOYSA-N | |
methylsulfanylmethane | |
1068 | |
62.13 | |
Extra Pure | |
Methyl sulfide |
-98.0°C | |
0.8460g/mL | |
−48°C | |
0.2% max. Methylmercaptan (GC) | |
(CH3)2S | |
MFCD00008562 | |
02,156; 04,190; 05,260; 15,88; 16,84; 17,198 | |
0.846 | |
Solubility in water: 2g/L (20°C). Other solubilities: soluble in alcohol and ether | |
CSC | |
62.13 | |
CHEBI:17437 | |
99+% | |
Liquid |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-846-2
- Methyl sulfide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only